Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Serine Protease Inhibitor Cocktail I

Supplier: Alfa Aesar
J64695-LQ J64695-LQ
AAJ64695-LQEA 383.85 USD InStock
AAJ64695-LQ CAAAJ64695-LQ
Serine Protease Inhibitor Cocktail I
Cell Culture Supplements and Additives
Lyophilized solid

Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized solid
Sensitivity: Hygroscopic
Appearance: White
UN No.:UN3261

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty