Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Pituitary Extract, BT-215

Supplier: Alfa Aesar
J64417-MF J64417-MF
AAJ64417-MFEA 120.96 USD InStock
AAJ64417-MF CAAAJ64417-MF
Bovine Pituitary Extract, BT-215
Proteins and Peptides
Lyophilized powder

Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
MDL No.: MFCD00163156

Applications: Bovine Pituitary Extract, BPE, is an extract prepared from bovine pituitary glands. BPE also known as keratinocyte growth supplement is used in conjunction with other growth factors, hormones and media to reduce or eliminate serum in the culturing of some epithelial cell types. BPE has been effectively utilized in culturing normal human epidermal keratinocytes (Tsao, M.C., et al, J. Cell Physiol. 110: 219-229, 1982) and other epithelial cells. BPE is membrane filtered and packaged as a lyophilized solid.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty