11-Azido-3,6,9-trioxaundecan-1-amine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane11-Amino-3,6,9-trioxaundecanyl AzideAmino-PEG3-AzideAzido-PEG3-Amine

J64308-MC J64308-MC
AAJ64308-MCEA 298.27 USD
AAJ64308-MC CAAAJ64308-MC
11-Azido-3,6,9-trioxaundecan-1-amine
11-Azido-3,6,9-trioxaundecan-1-amine

Viscous liquid

Formula: C₈H₁₈N₄O₃
MW: 218.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00269874
CAS Number: 134179-38-7
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR