Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxyanthranilic acid

Supplier: Alfa Aesar
Warning
Synonyms: 2-Amino-3-methoxybenzoic acid

J64244-09 J64244-06 J64244-14 J64244-09 J64244-14 J64244-06
AAJ64244-09EA 273.02 USD InStock
AAJ64244-09 CAAAJ64244-06 AAJ64244-14 CAAAJ64244-09 CAAAJ64244-14 AAJ64244-06
3-Methoxyanthranilic acid
3-Methoxyanthranilic acid
Powder


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 169-170°C
Storage Temperature: Ambient
MDL Number: MFCD00075178
CAS Number: 3177-80-8
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty