Cortisone

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 17alpha,21-Dihydroxy-4-pregnene-3,11,20-trione17alpha-Hydroxy-11-dehydrocorticosterone

J64182-03 J64182-06 J64182-03 J64182-06
AAJ64182-03EA 58.11 USD
CAAAJ64182-06 AAJ64182-03 AAJ64182-06 CAAAJ64182-03
Cortisone
Cortisone

A steroid and immunosuppressant


Powder

Formula: C₂₁H₂₈O₅
MW: 360.44
Melting Pt: 223…228 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003610
CAS Number: 53-06-5
EINECS: 200-162-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR