Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cellulase, chromatographically purified, T. reesei

Supplier: Alfa Aesar
J64019-MD J64019-03 J64019-03 J64019-MD
AAJ64019-MDEA 72.89 USD InStock
AAJ64019-MD CAAAJ64019-03 AAJ64019-03 CAAAJ64019-MD
Cellulase, chromatographically purified, T. reesei
Enzymes
Lyophilized powder

CAS: 9012-54-8
MDL: MFCD00081510

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty