Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-cAMP Rabbit 0 Antibody

Supplier: Alfa Aesar
J64001-KX J64001-KX
AAJ64001-KXEA 66.35 USD InStock
CAAAJ64001-KX AAJ64001-KX
Anti-cAMP Rabbit 0 Antibody
Antibodies
MDL No.: MFCD00162208

Type:
Antigen: cAMP
Clonality: 0
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty