Anisomycin 97%

Supplier: Thermo Scientific Chemicals
Danger

J62964-MF J62964-MF
AAJ62964-MFEA 277.33 USD
CAAAJ62964-MF AAJ62964-MF
Anisomycin 97%
Anisomycin

A protein synthesis inhibitor. Causes apoptosis of PC12 Cells


Powder

Formula: C₁₄H₁₉NO₄
Melting Pt: 140…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077650
CAS Number: 22862-76-6
EINECS: 245-269-7
UN: 3462
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR