Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromomycin A3 ≥97%

Supplier: Alfa Aesar
Danger

J62927-MA J62927-MA
AAJ62927-MAEA 388.73 USD InStock
AAJ62927-MA CAAAJ62927-MA
Chromomycin A3 ≥97%
Chromomycin A3
Chromomycin A3 is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye.

  • Yellow powder

Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate "G-C" box, induced by oxidative stress or DNA damage. Furthermore, by inhibiting transcription, Chromomycin A3 and its structural analogs can inhibit protein biosynthesis.

Applications: An inhibitor of DNA and RNA polymerases. Serves as a fluorescent DNA stain

Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.


Formula: C₅₇H₈₂O₂₆
MW: 1183,30 g/mol
Melting Pt: 185 °C
Storage Temperature: Freezer
MDL Number: MFCD00043151
CAS Number: 7059-24-7
EINECS: 230-348-0
UN: 3462
ADR: ,
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty