Zymosan A (from S. cerevisiae)

Supplier: Thermo Scientific Chemicals

Synonyms: Zymosan

J62876-MD J62876-03 J62876-03 J62876-MD
AAJ62876-MDEA 48.77 USD
AAJ62876-MD CAAAJ62876-03 AAJ62876-03 CAAAJ62876-MD
Zymosan A (from S. cerevisiae)
Zymosan A

An insoluble preparation of yeast cell, has been shown to activate macrophages via toll-like receptor 2

Storage Temperature: Ambient
MDL Number: MFCD00082157
CAS Number: 58856-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR