Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Galantamine hydrobromide

Supplier: Alfa Aesar
Danger

J62482-MF J62482-03 J62482-03 J62482-MF
AAJ62482-MFEA 187.94 USD InStock
AAJ62482-MF CAAAJ62482-03 AAJ62482-03 CAAAJ62482-MF
Galantamine hydrobromide
Galantamine hydrobromide
  • Soluble in water or DMSO

Powder

CAS No.: 1953-04-4
Molecular Formula: C17H21NO3.HBr
Formula Weight: 368.27
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water or DMSO
UN No.: 2811
MDL No.: MFCD00067672

Applications: Centrally active and long-acting acetylcholinesterase inhibitor


Formula: C₁₇H₂₁NO₃·HBr
MW: 368,27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067672
CAS Number: 1953-04-4
EINECS: 217-780-5
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty