Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylazine hydrochloride

Supplier: Alfa Aesar
Danger
Synonyms: N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride2-[(2,6-Dimethylphenyl)amino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride

J62394-06 J62394-03 J62394-03 J62394-06 J62394-14 J62394-14
AAJ62394-06EA 146.05 USD InStock
AAJ62394-06 CAAAJ62394-03 AAJ62394-03 CAAAJ62394-06 CAAAJ62394-14 AAJ62394-14
Xylazine hydrochloride
Xylazine hydrochloride
Powder

Applications: Agonist at the alpha-2 adrenergic receptor


Formula: C₁₂H₁₆N₂S
MW: 256.79 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058196
CAS Number: 23076-35-9
EINECS: 245-417-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,10080
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty