Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Valinomycin ≥90%

Supplier: Alfa Aesar
Danger

J62312-MC J62312-MC
AAJ62312-MCEA 294.76 USD InStock
AAJ62312-MC CAAAJ62312-MC
Valinomycin ≥90%
Valinomycin
Valinomycin is used as a potassium ionophore shown to induce cell death

  • White crystalline powder

Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

Applications: Potassium-selective ionophoric cyclodepsipeptide; inhibitor of mitochondrial action

Soluble in DMSO at 10mg/mL. Insoluble in water

Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.


Formula: C₅₄H₉₀O₁₈N₆
MW: 1111.34 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005114
CAS Number: 2001-95-8
EINECS: 217-896-6
UN: 2811
ADR: ,
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty