Josamycin ≥98%

Supplier: Thermo Scientific Chemicals

J62245-MF J62245-MF
AAJ62245-MFEA 110.56 USD
AAJ62245-MF CAAAJ62245-MF
Josamycin ≥98%
Josamycin

Josamycin is used as a macrolide antibiotic effective against a variety of pathogens.


White to pale yellow powder


Applications: Macrolide antibiotic


Soluble in ethanol


Caution: Protect from light and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Formula: C₄₂H₆₉NO₁₅
Storage Temperature: Refrigerator
MDL Number: MFCD00211043
CAS Number: 16846-24-5
EINECS: 240-871-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR