Phenolphthalein 2% in ethanol 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1(3H)-Isobenzofuranone, 3,3-bis(4-hydroxyphenyl)-
J61785-AE
J61785-AU
AAJ61785-AEEA
31.99
USD
AAJ61785-AE
AAJ61785-AU
Phenolphthalein 2% in ethanol 95%
Phenolphthalein
pH indicator
Liquid
Formula:
C₂₀H₁₄O₄ MW: 318.33 g/mol Flash Pt: 17 °C(63 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 UN: 1987 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...