Haloperidol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone

J61688-06 J61688-06
AAJ61688-06EA 114.93 USD
AAJ61688-06 CAAAJ61688-06
Haloperidol
Haloperidol

An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats


Powder

Formula: C₂₁H₂₃ClFNO₂
MW: 375.87 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00051423
CAS Number: 52-86-8
EINECS: 200-155-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR