Monensin monosodium salt ≥90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Monensin, Sodium SaltMonensin sodium salt

J61669-03 J61669-03
AAJ61669-03EA 96.79 USD
CAAAJ61669-03 AAJ61669-03
Monensin monosodium salt ≥90%
Monensin monosodium salt

Antibiotic that functions as an sodium ionophore. Induces apoptosis in HL-60 cells


Powder

Formula: C₃₆H₆₁NaO₁₁
MW: 692.85 g/mol
Melting Pt: 267…269 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077826
CAS Number: 22373-78-0
EINECS: 244-941-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR