Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium chloride 0.5 mol/l in aqueous solution

Supplier: Alfa Aesar
Warning
Synonyms: Aluminum chloride

J61582-AD J61582-AE J61582-AE J61582-AD
AAJ61582-ADEA 97.24 USD InStock
AAJ61582-AD CAAAJ61582-AE AAJ61582-AE CAAAJ61582-AD
Aluminium chloride 0.5 mol/l in aqueous solution
Aluminium chloride
Liquid


Formula: AlCl₃
MW: 133,34 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 180 to 181 °C
Density: liquid 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 2581
ADR: 8,III
Merck Index: 13,00335
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty