Gelatine Type A

Supplier: Thermo Scientific
J61548-36 J61548-A1 J61548-22
AAJ61548-36EA 138.75 USD
AAJ61548-36 AAJ61548-A1 AAJ61548-22
Gelatine Type A
Proteins and Peptides
CAS No.: 9000-70-8
Physical Form: Powder
Appearance: Off-white to yellow
MDL No.: MFCD00081638
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR