Verapamil hydrochloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride
J61535-03
J61535-03
AAJ61535-03EA
87.21
USD
AAJ61535-03
CAAAJ61535-03
Verapamil hydrochloride 99+%
Verapamil hydrochloride
A calcium channel blocker with a chemopreventive character
Powder
Formula:
C₂₇H₃₈N₂O₄ MW: 491.07 g/mol Melting Pt: 138…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055208 CAS Number: 152-11-4 EINECS: 205-800-5 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...