Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Physostigmine ≥98.5% (by HPLC), white to off-white powder

Supplier: Alfa Aesar
Danger
Synonyms: Eserine

J61477-MC J61477-MC
AAJ61477-MCEA 38.49 USD InStock
CAAAJ61477-MC AAJ61477-MC
Physostigmine ≥98.5% (by HPLC), white to off-white powder
Physostigmine
Eserine is a parasympathomimetic, specifically, a reversible cholinesterase inhibitor obtained from the calabar bean, used to treat glaucoma and delayed gastric emptying.

This compound has been used as a positive control in a study investigating acetylcholinesterase inhibitors for potential antihelminthic drug targets. Additionally, this parasympathomimetic compound is reported to cross the blood-brain barrier. It is also used as a veterinary medication.


Formula: C₁₅H₂₁N₃O₂
MW: 275.35 g/mol
Melting Pt: 102-104°C
Storage Temperature: Ambient
MDL Number: MFCD00151090
CAS Number: 57-47-6
EINECS: 200-332-8
UN: 1544
ADR: 6.1,I
Order Now
Specification Test Results
Assay (HPLC) Min. 98.5%
Specific rotation (C=2 in chloroform, 20 °C) −76 to −80°

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty