1,4-Butanediol dimethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Busulfan
J61348-06
J61348-06
AAJ61348-06EA
26.77
USD
CAAAJ61348-06
AAJ61348-06
1,4-Butanediol dimethanesulfonate 98%
Busulfan
A cell cycle non-specific alkylating antineoplastic agent
Crystalline powder
Formula:
C₆H₁₄O₆S₂ MW: 246.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007562 CAS Number: 55-98-1 EINECS: 200-250-2 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...