N-(3-Chloro-2-methylphenyl)anthranilic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-[(3-Chloro-2-methylphenyl)amino]benzoic Acid2-(3-Chloro-o-toluidino)benzoic AcidTolfenamic Acid

J61256-09 J61256-06 J61256-14 J61256-06 J61256-14 J61256-09
AAJ61256-06EA 87.43 USD
AAJ61256-06 AAJ61256-14 CAAAJ61256-09 CAAAJ61256-14 CAAAJ61256-06 AAJ61256-09
N-(3-Chloro-2-methylphenyl)anthranilic acid 99+%
N-(3-Chloro-2-methylphenyl)anthranilic acid

NSAID that activates calcium-activated potassium channels


Crystalline powder

Formula: C₁₄H₁₂ClNO₂
MW: 261.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133865
CAS Number: 13710-19-5
EINECS: 237-264-3
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR