Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluophenazine ≥98%

Supplier: Alfa Aesar
Danger

J61179-06 J61179-06 J61179-03 J61179-03
AAJ61179-06EA 287.94 USD InStock
AAJ61179-06 CAAAJ61179-06 AAJ61179-03 CAAAJ61179-03
Fluophenazine ≥98%
Fluophenazine
Powder

Applications: Dopamine D1 receptor antagonist


Formula: C₂₂H₂₆F₃N₃OS
MW: 437.525 g/mol
Melting Pt: 268-274°C
Storage Temperature: Ambient
MDL Number: MFCD00242759
CAS Number: 69-23-8
EINECS: 200-702-9
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty