Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbadox ≥98%

Supplier: Alfa Aesar
Danger

J60284-14 J60284-22 J60284-14 J60284-22
AAJ60284-14EA 94.78 USD InStock
AAJ60284-14 CAAAJ60284-22 CAAAJ60284-14 AAJ60284-22
Carbadox ≥98%
Carbadox
Yellow solid

Solid

Purity: 98+%
CAS No.: 6804-07-5
Molecular Formula: C11H10N4O4
Formula Weight: 262.22
Flash Point: 18°(64°F)
Physical Form: Solid
Appearance: Yellow
Melting Point: 239-240°
UN No.: 1325
MDL No.: MFCD00057293

Applications: Quinoxaline derivative. A broad spectrum antibiotic agent


Formula: C₁₁H₁₀N₄O₄
MW: 262.22 g/mol
Melting Pt: 239…240 °C
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00057293
CAS Number: 6804-07-5
UN: 1325
ADR: 4.1,II
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty