(+/-)-Thalidomide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(2,6-Dioxo-3-piperidinyl)phthalimide
J60271-MC
J60271-MC
AAJ60271-MCEA
73.84
USD
AAJ60271-MC
CAAAJ60271-MC
(+/-)-Thalidomide
(+/-)-Thalidomide
Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis
Powder
Formula:
C₁₃H₁₀N₂O₄ MW: 258.23 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00153873 CAS Number: 50-35-1 EINECS: 200-031-1 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...