Acriflavine hydrochloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,6-Diamino-10-methylacridinium chloride hydrochloride
J60048-14
J60048-14
AAJ60048-14EA
53.14
USD
AAJ60048-14
CAAAJ60048-14
Acriflavine hydrochloride
Acriflavine hydrochloride
An intercalating dye.
Powder
Formula:
C₂₇H₂₈Cl₄N₆ MW: 578.36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069039 CAS Number: 8063-24-9 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...