6-Bromoindole-3-carboxaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Bromo-1H-indole-3-carbaldehyde
, 6-Bromo-3-formylindole
H66756-03
H66756-06
AAH66756-03EA
100.99
USD
AAH66756-03
AAH66756-06
6-Bromoindole-3-carboxaldehyde 95%
6-Bromoindole-3-carboxaldehyde
Formula:
C₉H₆BrNO MW: 224.06 g/mol Melting Pt: 202…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792689 CAS Number: 17826-04-9 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...