4-Chloroindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-1H-indole

H66153-06 H66153-03
AAH66153-06EA 118.99 USD
AAH66153-06 AAH66153-03
4-Chloroindole 97%
4-Chloroindole
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 129…130 °C/4 mm
Density: 1.259
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005665
CAS Number: 25235-85-2
EINECS: 246-747-8
UN: 2810
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR