2,4-Diaminothiazole hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Thiazole-2,4-diamine hydrochloride
H65434-06
H65434-03
H65434-MD
AAH65434-03EA
121.99
USD
AAH65434-03
AAH65434-MD
AAH65434-06
2,4-Diaminothiazole hydrochloride 95%
2,4-Diaminothiazole hydrochloride
Formula:
C₃H₆ClN₃S MW: 151.612 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01438626 CAS Number: 72792-54-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...