Methyl-2-(bromomethyl)-3-nitrobenzoate 95%

Danger

Synonyms: Methyl 2-(bromomethyl)-3-nitrobenzoate

H65351-MD H65351-03
AAH65351-03EA 300.81 USD
AAH65351-03 AAH65351-MD
Methyl-2-(bromomethyl)-3-nitrobenzoate 95%
Methyl 2-(bromomethyl)-3-nitrobenzoate
Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114315
CAS Number: 98475-07-1
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR