1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 98%
|
Warning
|
Synonyms:
tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate
, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole
H65315-06
H65315-MD
H65315-03
AAH65315-06EA
301.79
USD
AAH65315-06
AAH65315-03
AAH65315-MD
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 98%
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole
Formula:
C₁₃H₂₀BrN₃O₂ MW: 330.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09831980 CAS Number: 877399-50-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...