2-Mercapto-4,5-dimethylthiazole 95%

Warning

Synonyms: 4,5-Dimethylthiazole-2(3H)-thione

H65085-06 H65085-MD H65085-03
AAH65085-MDEA 23.63 USD
AAH65085-MD AAH65085-06 AAH65085-03
2-Mercapto-4,5-dimethylthiazole 95%
2-Mercapto-4,5-dimethylthiazole
Formula: C₅H₇NS₂
MW: 145.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037895
CAS Number: 5351-51-9
EINECS: 226-327-0

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR