3-Amino-4-fluoro-1H-indazole 97%

Warning

Synonyms: 4-Fluoro-1H-indazol-3-amine

H64990-03 H64990-MD H64990-06
AAH64990-03EA 97.56 USD
AAH64990-03 AAH64990-MD AAH64990-06
3-Amino-4-fluoro-1H-indazole 97%
3-Amino-4-fluoro-1H-indazole
Formula: C₇H₆FN₃
MW: 151.14 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00119934
CAS Number: 404827-78-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR