2-Fluoro-4-iodo-5-methylpyridine 95%

Warning

Synonyms: 2-Fluoro-4-iodo-5-picoline

H64983-03 H64983-06 H64983-MD
AAH64983-03EA 184.38 USD
AAH64983-03 AAH64983-MD AAH64983-06
2-Fluoro-4-iodo-5-methylpyridine 95%
2-Fluoro-4-iodo-5-methylpyridine
Formula: C₆H₅FIN
MW: 237.02 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD03095297
CAS Number: 153034-94-7

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR