Dimethyl-2,3-pyridinedicarboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethyl pyridine-2,3-dicarboxylate
H64939-06
H64939-22
H64939-14
AAH64939-14EA
92.89
USD
AAH64939-14
AAH64939-22
AAH64939-06
Dimethyl-2,3-pyridinedicarboxylate 95%
Dimethyl 2,3-Pyridinedicarboxylate
Formula:
C₉H₉NO₄ MW: 195.17 g/mol Melting Pt: 53…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00186549 CAS Number: 605-38-9 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...