10-Phenyl-9-anthraceneboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(10-Phenylanthracen-9-yl)boronic acid
H64911-MD
H64911-06
H64911-03
AAH64911-MDEA
33.99
USD
AAH64911-MD
AAH64911-06
AAH64911-03
10-Phenyl-9-anthraceneboronic acid 98%
10-Phenyl-9-anthraceneboronic acid
Formula:
C₂₀H₁₅BO₂ MW: 298.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11111989 CAS Number: 334658-75-2 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...