4-Bromo-1-naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1-naphthalenecarboxylic acid

H64820-03 H64820-06
AAH64820-03EA 46.39 USD
AAH64820-03 AAH64820-06
4-Bromo-1-naphthoic acid 98%
4-Bromo-1-naphthoic acid
Formula: C₁₁H₇BrO₂
MW: 251.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094221
CAS Number: 16650-55-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR