Methyl-4-methylsalicylate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-hydroxy-4-methylbenzoate
H64793-14
H64793-06
AAH64793-06EA
50.49
USD
AAH64793-06
AAH64793-14
Methyl-4-methylsalicylate 98+%
Methyl 4-methylsalicylate
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 242° lit. Melting Pt: 27…28 °C Density: 1.147 Storage Temperature: Ambient |
MDL Number:
MFCD00020130 CAS Number: 4670-56-8 EINECS: 225-117-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...