3-chloro-4-hydroxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloropyridin-4-ol

H64783-03 H64783-06 H64783-MD
AAH64783-03EA 50.79 USD
AAH64783-03 AAH64783-06 AAH64783-MD
3-chloro-4-hydroxypyridine 98%
3-chloro-4-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.543 g/mol
Melting Pt: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD04114248
CAS Number: 89284-20-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR