9,9-Dimethylfluoren-2-boronic acid 98%

Warning

Synonyms: (9,9-Dimethyl-9H-fluoren-2-yl)boronic acid

H64777-06 H64777-03 H64777-MD
AAH64777-MDEA 23.24 USD
AAH64777-MD AAH64777-06 AAH64777-03
9,9-Dimethylfluoren-2-boronic acid 98%
9,9-Dimethylfluoren-2-boronic acid
Formula: C₁₅H₁₅BO₂
MW: 238.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704227
CAS Number: 333432-28-3

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR