3-Amino-2-fluoro-4-methylpyridine 98%

Warning

Synonyms: 2-Fluoro-4-methylpyridin-3-amine

H64758-03 H64758-MD H64758-06
AAH64758-03EA 47.24 USD
AAH64758-03 AAH64758-06 AAH64758-MD
3-Amino-2-fluoro-4-methylpyridine 98%
3-Amino-2-fluoro-4-methylpyridine
Formula: C₆H₇FN₂
MW: 126.13 g/mol
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD04112556
CAS Number: 173435-32-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR