4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride

H64755-MD H64755-03
AAH64755-03EA 304.99 USD
AAH64755-03 AAH64755-MD
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride
Formula: C6H10ClN3
MW: 159.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846200
CAS Number: 879668-17-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR