2,4-Dibromobenzyl alcohol 98%

Warning

Synonyms: (2,4-Dibromophenyl)methanol

H64741-03 H64741-MD H64741-06
AAH64741-03EA 92.57 USD
AAH64741-03 AAH64741-06 AAH64741-MD
2,4-Dibromobenzyl alcohol 98%
2,4-Dibromobenzyl alcohol
Formula: C₇H₆Br₂O
MW: 265.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09996906
CAS Number: 666747-06-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR