(3-(Pyridin-3-yl)phenyl)boronic acid 98%

Warning

Synonyms: 3-(3-Pyridyl)benzeneboronic acid

H64739-03 H64739-MD H64739-06
AAH64739-MDEA 26.74 USD
AAH64739-MD AAH64739-06 AAH64739-03
(3-(Pyridin-3-yl)phenyl)boronic acid 98%
(3-(Pyridin-3-yl)phenyl)boronic acid
Formula: C₁₁H₁₀BNO₂
MW: 199.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658918
CAS Number: 351422-72-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR