1-N-Cbz-pyrrolidine-3-carboxylic acid 97%

Warning

Synonyms: 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid1-N-Cbz-Pyrrolidine-3-carboxylic acid

H64728-03 H64728-06 H64728-MD
AAH64728-MDEA 19.75 USD
AAH64728-MD AAH64728-06 AAH64728-03
1-N-Cbz-pyrrolidine-3-carboxylic acid 97%
1-N-Cbz-pyrrolidine-3-carboxylic acid
Formula: C13H15NO4
MW: 249.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179036
CAS Number: 188527-21-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR