3-Fluoro-2-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64705-MD H64705-06 H64705-03
AAH64705-03EA 95.09 USD
AAH64705-03 AAH64705-06 AAH64705-MD
3-Fluoro-2-methylpyridine 98%
3-Fluoro-2-methylpyridine
Formula: C₆H₆FN
Storage Temperature: Ambient
MDL Number: MFCD08705710
CAS Number: 15931-15-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR