4-Bromo-9,9-dimethyl-9H-fluorene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-9,9-dimethylfluorene

H64639-03 H64639-06 H64639-MD
AAH64639-03EA 57.83 USD
AAH64639-03 AAH64639-06 AAH64639-MD
4-Bromo-9,9-dimethyl-9H-fluorene 98%
4-Bromo-9,9-dimethyl-9H-fluorene
Formula: C₁₅H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD23099415
CAS Number: 942615-32-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR