4-Bromo-2,6-difluorophenylboronic acid 98%

Warning

Synonyms: 4-Bromo-2,6-difluorobenzeneboronic acid

H64609-06 H64609-03
AAH64609-06EA 145.41 USD
AAH64609-06 AAH64609-03
4-Bromo-2,6-difluorophenylboronic acid 98%
4-Bromo-2,6-difluorophenylboronic acid
Formula: C₆H₄BBrF₂O₂
MW: 236.807 g/mol
Melting Pt: 155…160 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD03095369
CAS Number: 352535-81-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR