1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 95%

Warning

Synonyms: 1-Bromo-2-[3-(trifluoromethyl)phenyl]ethane

H64601-MD H64601-03 H64601-06
AAH64601-03EA 82.67 USD
AAH64601-03 AAH64601-06 AAH64601-MD
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 95%
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Formula: C₉H₈BrF₃
MW: 253.062 g/mol
Boiling Pt: 92…94 °C/12 mm
Density: 1.495
Storage Temperature: Ambient
MDL Number: MFCD03092900
CAS Number: 1997-80-4
UN: 3082
ADR: "9",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR